accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 33931-42-9

IUPAC Name: N,N,N',N'-tetraethylpropanediamide
Synonyms: N,N,N',N'-tetraethylpropanediamide
Molecular Weight (g/mol): 214.31
Molecular Formula: C11H22N2O2
InChi Key: RTHWPWJSSJEQNU-UHFFFAOYSA-N
SMILES: CCN(CC)C(=O)CC(=O)N(CC)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results