Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,N',N'-Tetraethylmalonamide 97.0+%, TCI America™
SDP

Catalog No. T19665G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T19665G Supplier TCI America Supplier No. T19665G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 33931-42-9
Molecular Formula C11H22N2O2
Molecular Weight (g/mol) 214.309
MDL Number MFCD06797171
InChI Key RTHWPWJSSJEQNU-UHFFFAOYSA-N
PubChem CID 551770
IUPAC Name N,N,N',N'-tetraethylpropanediamide
SMILES CCN(CC)C(=O)CC(=O)N(CC)CC

Specifications

Boiling Point 162°C
Quantity 5 g
Formula Weight 214.31
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N,N′,N′-Tetraethylmalonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.