accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 33941-15-0

IUPAC Name: 1,4,7,10,13-pentaoxa-16-azacyclooctadecan-16-ium
Synonyms: 1,4,7,10,13-pentaoxa-16-azacyclooctadecan-16-ium
Molecular Weight (g/mol): 264.34
Molecular Formula: C12H26NO5
InChi Key: NBXKUSNBCPPKRA-UHFFFAOYSA-O
SMILES: C1COCCOCCOCCOCCOCC[NH2+]1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (3)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)