accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 33953-37-6

IUPAC Name: N-(piperidin-4-yl)benzamide
Synonyms: N-(piperidin-4-yl)benzamide
Molecular Weight (g/mol): 204.27
Molecular Formula: C12H16N2O
InChi Key: JMQDNLCNCDSHNC-UHFFFAOYSA-N
SMILES: O=C(NC1CCNCC1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results