CAS RN 3396-11-0
Cesium acetate, 99.9% (metals basis)
CAS: 3396-11-0 Molecular Formula: C2H3CsO2 Molecular Weight (g/mol): 191.949 MDL Number: MFCD00013056 InChI Key: ZOAIGCHJWKDIPJ-UHFFFAOYSA-M Synonym: cesium acetate,acetic acid, cesium salt,caesium acetate,acetic acid, cesium salt 1:1,acetic acid, cesiumsalt,acetic acid cesium salt,caesium 1+ ion acetate,cesiumacetate,acetic acid cesium,caesium 1+ acetate ion PubChem CID: 5152919 IUPAC Name: cesium;acetate SMILES: CC(=O)[O-].[Cs+]
Cesium acetate, 99%, for analysis
CAS: 3396-11-0 Molecular Formula: C2H3CsO2 Molecular Weight (g/mol): 191.96 MDL Number: MFCD00013056 InChI Key: ZOAIGCHJWKDIPJ-UHFFFAOYSA-M Synonym: cesium acetate,acetic acid, cesium salt,caesium acetate,acetic acid, cesium salt 1:1,acetic acid, cesiumsalt,acetic acid cesium salt,caesium 1+ ion acetate,cesiumacetate,acetic acid cesium,caesium 1+ acetate ion PubChem CID: 5152919 IUPAC Name: cesium;acetate SMILES: CC(=O)[O-].[Cs+]
Cesium Acetate, 99.9%, MP Biomedicals
CAS: 3396-11-0 Molecular Formula: C2H3CsO2 Molecular Weight (g/mol): 191.949 InChI Key: ZOAIGCHJWKDIPJ-UHFFFAOYSA-M Synonym: cesium acetate,acetic acid, cesium salt,caesium acetate,acetic acid, cesium salt 1:1,acetic acid, cesiumsalt,acetic acid cesium salt,caesium 1+ ion acetate,cesiumacetate,acetic acid cesium,caesium 1+ acetate ion PubChem CID: 5152919 IUPAC Name: cesium;acetate SMILES: CC(=O)[O-].[Cs+]
Cesium acetate, 99.998% (metals basis)
CAS: 3396-11-0 Molecular Formula: C2H3CsO2 Molecular Weight (g/mol): 191.949 MDL Number: MFCD00013056 InChI Key: ZOAIGCHJWKDIPJ-UHFFFAOYSA-M Synonym: cesium acetate,acetic acid, cesium salt,caesium acetate,acetic acid, cesium salt 1:1,acetic acid, cesiumsalt,acetic acid cesium salt,caesium 1+ ion acetate,cesiumacetate,acetic acid cesium,caesium 1+ acetate ion PubChem CID: 5152919 IUPAC Name: cesium;acetate SMILES: CC(=O)[O-].[Cs+]
Cesium acetate, 97%, pure
CAS: 3396-11-0 Molecular Formula: C2H3CsO2 Molecular Weight (g/mol): 191.96 MDL Number: MFCD00013056 InChI Key: ZOAIGCHJWKDIPJ-UHFFFAOYSA-M Synonym: cesium acetate,acetic acid, cesium salt,caesium acetate,acetic acid, cesium salt 1:1,acetic acid, cesiumsalt,acetic acid cesium salt,caesium 1+ ion acetate,cesiumacetate,acetic acid cesium,caesium 1+ acetate ion PubChem CID: 5152919 IUPAC Name: cesium;acetate SMILES: CC(=O)[O-].[Cs+]
Cesium acetate, 99% (metals basis)
CAS: 3396-11-0 Molecular Formula: C2H3CsO2 Molecular Weight (g/mol): 191.949 MDL Number: MFCD00013056 InChI Key: ZOAIGCHJWKDIPJ-UHFFFAOYSA-M Synonym: cesium acetate,acetic acid, cesium salt,caesium acetate,acetic acid, cesium salt 1:1,acetic acid, cesiumsalt,acetic acid cesium salt,caesium 1+ ion acetate,cesiumacetate,acetic acid cesium,caesium 1+ acetate ion PubChem CID: 5152919 IUPAC Name: cesium;acetate SMILES: CC(=O)[O-].[Cs+]
Cesium Acetate 98.0+%, TCI America™
CAS: 3396-11-0 Molecular Formula: C2H3CsO2 Molecular Weight (g/mol): 191.949 MDL Number: MFCD00013056 InChI Key: ZOAIGCHJWKDIPJ-UHFFFAOYSA-M Synonym: cesium acetate,acetic acid, cesium salt,caesium acetate,acetic acid, cesium salt 1:1,acetic acid, cesiumsalt,acetic acid cesium salt,caesium 1+ ion acetate,cesiumacetate,acetic acid cesium,caesium 1+ acetate ion PubChem CID: 5152919 IUPAC Name: cesium;acetate SMILES: CC(=O)[O-].[Cs+]
Sigma Aldrich Cesium acetate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Cesium acetate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Cesium Acetate, 99.9%, Optical Grade, MP Biomedicals™
CAS: 3396-11-0 Molecular Formula: C2H3CsO2 Molecular Weight (g/mol): 191.949 InChI Key: ZOAIGCHJWKDIPJ-UHFFFAOYSA-M Synonym: cesium acetate,acetic acid, cesium salt,caesium acetate,acetic acid, cesium salt 1:1,acetic acid, cesiumsalt,acetic acid cesium salt,caesium 1+ ion acetate,cesiumacetate,acetic acid cesium,caesium 1+ acetate ion PubChem CID: 5152919 IUPAC Name: cesium;acetate SMILES: CC(=O)[O-].[Cs+]