Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium Acetate, 99.9%, Optical Grade, MP Biomedicals™

Catalog No. ICN10131925 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10131925 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10131925 Supplier MP Biomedicals, Inc Supplier No. 0210131925

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3396-11-0
Molecular Formula C2H3CsO2
Molecular Weight (g/mol) 191.949
InChI Key ZOAIGCHJWKDIPJ-UHFFFAOYSA-M
Synonym cesium acetate, acetic acid, cesium salt, caesium acetate, acetic acid, cesium salt 1:1, acetic acid, cesiumsalt, acetic acid cesium salt, caesium 1+ ion acetate, cesiumacetate, acetic acid cesium, caesium 1+ acetate ion
PubChem CID 5152919
IUPAC Name cesium;acetate
SMILES CC(=O)[O-].[Cs+]

Specifications

Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.9%
Formula Weight 191.9
Grade Optical
Chemical Name or Material Cesium Acetate
Recommended Storage Store at room temperature.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.