CAS RN 3396-99-4
IUPAC Name:
2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
Synonyms:
methyl-α-D-mannoside methyl mannoside mannoside, methyl
Molecular Weight (g/mol):
194.18
Molecular Formula:
C7H14O6
InChi Key:
HOVAGTYPODGVJG-UHFFFAOYNA-N
SMILES:
COC1OC(CO)C(O)C(O)C1O
1
–
1
of
1
results
1
Promotions Available
Methyl-α-D-galactopyranoside, 98+%
Methyl-α-D-galactopyranoside, 98+%
CAS: 3396-99-4 MDL Number: MFCD00064085 PubChem CID: 76935 ChEBI: CHEBI:55507