Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl-α-D-galactopyranoside, 98+%

Catalog No. AC337090500
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC337090500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC337090500 Supplier Thermo Scientific Chemicals Supplier No. 337090500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 3396-99-4 | C7H14O6 | 194.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3396-99-4
MDL Number MFCD00064085
PubChem CID 76935
ChEBI CHEBI:55507

Specifications

Melting Point 116.0°C to 117.0°C
Assay Percent Range 98% min. (ELSD) (HPLC)
Packaging Glass Bottle
Molecular Formula C7H14O6
Quantity 50 g
Specific Rotation Condition + 179.00 (20.00°C c=1,water)
Specific Rotation + 179.00
Synonym methyl alpha-d-galactopyranoside, alpha-methyl-d-galactoside, alpha-methylgalactoside, me-alpha-gal, methyl alpha-d-galactoside, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-methoxytetrahydro-2h-pyran-3,4,5-triol, methyl, a-d-galactopyranoside, methyl-galactopyranoside, methyl-alpha-d-galactopyranoside, alpha-methyl d-galactoside
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol
InChI Key HOVAGTYPODGVJG-PZRMXXKTSA-N
SMILES COC1C(C(C(C(O1)CO)O)O)O
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
Molecular Weight (g/mol) 194.18
Formula Weight 194.18
Percent Purity 98+%
Chemical Name or Material Methyl-α-D-galactopyranoside
Show More Show Less
EINECSNumber 222-251-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.