accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHOONOOO

CAS RN 3397-32-8

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 2-{[(benzyloxy)carbonyl]amino}-3-phenylpropanoate
Synonyms: 2,5-dioxopyrrolidin-1-yl 2-{[(benzyloxy)carbonyl]amino}-3-phenylpropanoate
Molecular Weight (g/mol): 396.4
Molecular Formula: C21H20N2O6
InChi Key: MOJNKVWQJKPXCT-UHFFFAOYNA-N
SMILES: O=C(NC(CC1=CC=CC=C1)C(=O)ON1C(=O)CCC1=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)