accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHFFF

CAS RN 340-06-7

IUPAC Name: 2,2,2-trifluoro-1-phenylethan-1-ol
Synonyms: 2,2,2-trifluoro-1-phenylethanol
Molecular Weight (g/mol): 176.14
Molecular Formula: C8H7F3O
InChi Key: VNOMEAQPOMDWSR-UHFFFAOYNA-N
SMILES: OC(C1=CC=CC=C1)C(F)(F)F
Boiling Point 
  • (2)
  • (1)
Quantity 
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)