Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,2'-Methylenebis[(4S)-4-Phenyl-2-Oxazoline]

Catalog No. 4164281G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-642-81G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 41-642-81G Supplier Sigma Aldrich Supplier No. 4164281G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

C2 symmetric ligand for enantioselective catalysis. Easily forms bidentate coordination complexes due to the strong affinity of the oxazoline nitrogen for various metals.

Specifications

CAS 340-06-7
Density 1.3 g/mL at 25°C
Boiling Point 73°C to 76°C (9 mmHg, lit.)
Molecular Formula C8H7F3O
Refractive Index n20/D 1.586
Linear Formula C6H5CH(CF3)OH
MDL Number MFCD00077845
Quantity 1 g
Synonym (S,S)-2,2'-Methylenebis(4-phenyl-2-oxazoline)
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.