accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3404-77-1

IUPAC Name: 3,3-dimethylhex-1-ene
Synonyms: 1-hexene, 3,3-dimethyl-
Molecular Weight (g/mol): 112.22
Molecular Formula: C8H16
InChi Key: RXYYKIMRVXDSFR-UHFFFAOYSA-N
SMILES: CCCC(C)(C)C=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results