Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3-Dimethyl-1-hexene 98.0+%, TCI America™
SDP

Catalog No. D12431ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12431ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D12431ML Supplier TCI America Supplier No. D12431ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3404-77-1
Molecular Formula C8H16
Molecular Weight (g/mol) 112.22
MDL Number MFCD00048727
InChI Key RXYYKIMRVXDSFR-UHFFFAOYSA-N
PubChem CID 137924
IUPAC Name 3,3-dimethylhex-1-ene
SMILES CCCC(C)(C)C=C

Specifications

Color Colorless
Boiling Point 104°C
Quantity 1 mL
UN Number 3295
Formula Weight 112.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,3-Dimethyl-1-hexene
EINECSNumber (2)-0024
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.