accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34040-64-7

IUPAC Name: methyl 4-(chloromethyl)benzoate
Synonyms: methyl 4-(chloromethyl)benzoate
Molecular Weight (g/mol): 184.62
Molecular Formula: C9H9ClO2
InChi Key: SATDLKYRVXFXRE-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(CCl)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results