Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-(Chloromethyl)benzoate 98.0+%, TCI America™
SDP

Catalog No. C17635G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17635G 5 g
C176325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17635G Supplier TCI America Supplier No. C17635G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 34040-64-7
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.619
MDL Number MFCD00511224
InChI Key SATDLKYRVXFXRE-UHFFFAOYSA-N
Synonym methyl 4-chloromethyl benzoate, 4-chloromethyl benzoic acid methyl ester, methyl-4-chloromethylbenzoate, benzoic acid, 4-chloromethyl-, methyl ester, 4-methoxycarbonyl benzyl chloride, methyl alpha-chloro-p-toluate, methyl p-chloromethyl benzoate, p-methoxycarbonyl benzyl chloride, methyl a-chloro-p-toluate, p-toluic acid, .alpha.-chloro-, methyl ester
PubChem CID 36664
IUPAC Name methyl 4-(chloromethyl)benzoate
SMILES COC(=O)C1=CC=C(C=C1)CCl

Specifications

Melting Point 39°C
Color White-Yellow
Boiling Point 165°C
Quantity 5 g
UN Number 3261
Formula Weight 184.62
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-(Chloromethyl)benzoate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.