accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3CH3

CAS RN 3405-42-3

IUPAC Name: methyldipropylamine
Synonyms: methyldipropylamine
Molecular Weight (g/mol): 115.22
Molecular Formula: C7H17N
InChi Key: UVBMZKBIZUWTLV-UHFFFAOYSA-N
SMILES: CCCN(C)CCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results