accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOOH2N

CAS RN 34079-31-7

IUPAC Name: benzyl 2-carbamoylpyrrolidine-1-carboxylate
Synonyms: benzyl 2-carbamoylpyrrolidine-1-carboxylate
Molecular Weight (g/mol): 248.28
Molecular Formula: C13H16N2O3
InChi Key: ZCGHEBMEQXMRQL-UHFFFAOYNA-N
SMILES: NC(=O)C1CCCN1C(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)