Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-L-prolinamide 98.0+%, TCI America™
SDP

Catalog No. C30055G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30055G 5 g
C300525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30055G Supplier TCI America Supplier No. C30055G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34079-31-7
Molecular Formula C13H16N2O3
Molecular Weight (g/mol) 248.282
MDL Number MFCD00020830
InChI Key ZCGHEBMEQXMRQL-NSHDSACASA-N
Synonym z-pro-nh2, cbz-l-prolinamide, s-1-n-cbz-prolinamide, s-benzyl 2-carbamoylpyrrolidine-1-carboxylate, cbz-pro-nh2, z-l-prolinamide, z-l-proline amide, carbobenzyloxy-l-prolinamide, benzyl 2s-2-carbamoylpyrrolidine-1-carboxylate, benzyloxycarbonyl-l-proline amide
PubChem CID 6950961
IUPAC Name benzyl (2S)-2-carbamoylpyrrolidine-1-carboxylate
SMILES C1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)N

Specifications

Melting Point 92°C
Color White-Yellow
Quantity 5 g
Formula Weight 248.28
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-L-prolinamide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.