accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCH3SNOH3C

CAS RN 341028-37-3

IUPAC Name: 4,5-dimethyl-3-(2-oxo-2-phenylethyl)-1,3-thiazol-3-ium chloride
Synonyms: alagebrium alagebrium chloride
Molecular Weight (g/mol): 267.77
Molecular Formula: C13H14ClNOS
InChi Key: MKOMESMZHZNBIZ-UHFFFAOYSA-M
SMILES: [Cl-].CC1=C(C)[N+](CC(=O)C2=CC=CC=C2)=CS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results