Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alagebrium Chloride 98.0+%, TCI America™
SDP

Catalog No. A3166200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A3166200MG 200 mg
A31661G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A3166200MG Supplier TCI America Supplier No. A3166200MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 341028-37-3
Molecular Formula C13H14ClNOS
Molecular Weight (g/mol) 267.771
MDL Number MFCD09027398
InChI Key MKOMESMZHZNBIZ-UHFFFAOYSA-M
Synonym 4,5-Dimethyl-3-(2-oxo-2-phenylethyl)thiazolium Chloride
PubChem CID 216306
IUPAC Name 2-(4,5-dimethyl-1,3-thiazol-3-ium-3-yl)-1-phenylethanone;chloride
SMILES CC1=C(SC=[N+]1CC(=O)C2=CC=CC=C2)C.[Cl-]

Specifications

Melting Point 212°C
Color White-Yellow
Quantity 200 mg
Formula Weight 267.77
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Alagebrium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.