accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NH2

CAS RN 34124-14-6

IUPAC Name: 2-[2-(2-aminophenyl)ethyl]aniline
Synonyms: 2-[2-(2-aminophenyl)ethyl]aniline
Molecular Weight (g/mol): 212.3
Molecular Formula: C14H16N2
InChi Key: ZYHQGITXIJDDKC-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1CCC1=CC=CC=C1N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results