Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Ethylenedianiline, TCI America™
SDP

Catalog No. E0472500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0472500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E0472500G Supplier TCI America Supplier No. E0472500G

missing translation for 'validMainframeMsg'

Specifications

CAS 34124-14-6
Melting Point 74°C
Molecular Formula C14H16N2
MDL Number MFCD00127990
Quantity 500 g
Synonym alpha,alpha′-Bi-o-toluidine, 2,2′-Diaminobibenzyl, 2,2′-Diamino-1,2-diphenylethane
InChI Key ZYHQGITXIJDDKC-UHFFFAOYSA-N
SMILES NC1=CC=CC=C1CCC1=CC=CC=C1N
IUPAC Name 2-[2-(2-aminophenyl)ethyl]aniline
Molecular Weight (g/mol) 212.30
PubChem CID 98690
Formula Weight 212.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Ethylenedianiline
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.