accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOHNHOHNONHOO

CAS RN 34152-86-8

IUPAC Name: 2-[2-(2-{[(benzyloxy)carbonyl]amino}acetamido)acetamido]-3-methylbutanoic acid
Synonyms: 2-[2-(2-{[(benzyloxy)carbonyl]amino}acetamido)acetamido]-3-methylbutanoic acid
Molecular Weight (g/mol): 365.39
Molecular Formula: C17H23N3O6
InChi Key: OLTJNTLCZYWHKR-UHFFFAOYNA-N
SMILES: CC(C)C(NC(=O)CNC(=O)CNC(=O)OCC1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)