accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34156-69-9

IUPAC Name: manganese(2+) carbonate
Synonyms: manganese carbonate|manganese(II) carbonate
Molecular Weight (g/mol): 114.95
Molecular Formula: CMnO3
InChi Key: XMWCXZJXESXBBY-UHFFFAOYSA-L
SMILES: [Mn++].[O-]C([O-])=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)