CAS RN 3416-57-7
IUPAC Name:
2-(2-oxopropyl)-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms:
2-(2-oxopropyl)isoindole-1,3-dione
Molecular Weight (g/mol):
203.2
Molecular Formula:
C11H9NO3
InChi Key:
STMRGLKPBJVVEG-UHFFFAOYSA-N
SMILES:
CC(=O)CN1C(=O)C2=CC=CC=C2C1=O
1
–
1
of
1
results
1
Phthalimidoacetone 98.0+%, TCI America™
CAS: 3416-57-7 Molecular Formula: C11H9NO3 Molecular Weight (g/mol): 203.197 MDL Number: MFCD00088346 InChI Key: STMRGLKPBJVVEG-UHFFFAOYSA-N Synonym: N-Acetonylphthalimide, 1-(N-Phthalimidyl)-2-propanone PubChem CID: 235046 IUPAC Name: 2-(2-oxopropyl)isoindole-1,3-dione SMILES: CC(=O)CN1C(=O)C2=CC=CC=C2C1=O