Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phthalimidoacetone 98.0+%, TCI America™
SDP

Catalog No. P120325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P120325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P120325G Supplier TCI America Supplier No. P120325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3416-57-7
Molecular Formula C11H9NO3
Molecular Weight (g/mol) 203.197
MDL Number MFCD00088346
InChI Key STMRGLKPBJVVEG-UHFFFAOYSA-N
Synonym N-Acetonylphthalimide, 1-(N-Phthalimidyl)-2-propanone
PubChem CID 235046
IUPAC Name 2-(2-oxopropyl)isoindole-1,3-dione
SMILES CC(=O)CN1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 123°C
Color White-Yellow
Quantity 25 g
Formula Weight 203.20
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Phthalimidoacetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.