accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34183-22-7

IUPAC Name: hydrogen 1-{2-[2-hydroxy-3-(propylamino)propoxy]phenyl}-3-phenylpropan-1-one chloride
Synonyms: propafenone
Molecular Weight (g/mol): 377.91
Molecular Formula: C21H28ClNO3
InChi Key: XWIHRGFIPXWGEF-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CCCNCC(O)COC1=CC=CC=C1C(=O)CCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results