Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propafenone Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. P23011G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P23011G Supplier TCI America Supplier No. P23011G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34183-22-7
Molecular Formula C21H28ClNO3
Molecular Weight (g/mol) 377.909
MDL Number MFCD00079243
InChI Key XWIHRGFIPXWGEF-UHFFFAOYSA-N
Synonym 1-[2-[2-Hydroxy-3-(propylamino)propoxy]phenyl]-3-phenyl-1-propanone Hydrochloride
PubChem CID 36708
ChEBI CHEBI:8466
IUPAC Name 1-[2-[2-hydroxy-3-(propylamino)propoxy]phenyl]-3-phenylpropan-1-one;hydrochloride
SMILES CCCNCC(COC1=CC=CC=C1C(=O)CCC2=CC=CC=C2)O.Cl

Specifications

Melting Point 173°C
Color White
Quantity 1 g
Formula Weight 377.91
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Propafenone Hydrochloride
RTECSNumber UH2833000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.