accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNBrOH

CAS RN 34206-49-0

IUPAC Name: 5-bromo-3-hydroxy-1,2-dihydropyridin-2-one
Synonyms: 5-bromo-3-hydroxy-1H-pyridin-2-one
Molecular Weight (g/mol): 190
Molecular Formula: C5H4BrNO2
InChi Key: KXBXUHWCODHRHK-UHFFFAOYSA-N
SMILES: OC1=CC(Br)=CNC1=O
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results