accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO(S)(R)CH3(S)(S)CH3(S)O(S)H3CO(R)

CAS RN 34209-81-9

IUPAC Name: (1R,2S,6S,7S,10S,11R,14S)-6-acetyl-7,11-dimethyl-5-oxapentacyclo[8.8.0.0²,⁷.0⁴,⁶.0¹¹,¹⁶]octadec-16-en-14-yl acetate
Synonyms: (1R,2S,6S,7S,10S,11R,14S)-6-acetyl-7,11-dimethyl-5-oxapentacyclo[8.8.0.0²,⁷.0⁴,⁶.0¹¹,¹⁶]octadec-16-en-14-yl acetate
Molecular Weight (g/mol): 372.51
Molecular Formula: C23H32O4
InChi Key: YRLVTBXOGJMZNC-YEBVRWJESA-N
SMILES: CC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@@]4(C)[C@@H](CC5O[C@@]45C(C)=O)[C@@H]3CC=C2C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results