Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

16,17-Epoxypregnenolone Acetate 95.0+%, TCI America™
SDP

Catalog No. E00151G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E00151G 1 g
E0015100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E00151G Supplier TCI America Supplier No. E00151G

Chemical Identifiers

CAS 34209-81-9
Molecular Formula C23H32O4
Molecular Weight (g/mol) 372.505
MDL Number MFCD00135637
InChI Key YRLVTBXOGJMZNC-LEQCJQSISA-N
PubChem CID 134611731
SMILES CC(=O)C12C(O1)CC3C2(CCC4C3CC=C5C4(CCC(C5)OC(=O)C)C)C

Specifications

Melting Point 159°C
Color White
Quantity 1 g
Formula Weight 372.51
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 16,17-Epoxypregnenolone Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.