accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34257-63-1

IUPAC Name: [methyl(phenyl)amino]triphenylphosphanium iodide
Synonyms: [methyl(phenyl)amino]triphenylphosphanium iodide
Molecular Weight (g/mol): 495.34
Molecular Formula: C25H23INP
InChi Key: SAYKZWPCENNSDR-UHFFFAOYSA-M
SMILES: [I-].CN(C1=CC=CC=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results