Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(N-Methyl-N-phenylamino)triphenylphosphonium Iodide, TCI America™
SDP

Catalog No. M115110G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M115110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M115110G Supplier TCI America Supplier No. M115110G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34257-63-1
Molecular Formula C25H23INP
Molecular Weight (g/mol) 495.344
MDL Number MFCD00040544
InChI Key SAYKZWPCENNSDR-UHFFFAOYSA-M
Synonym N-Methylanilinotriphenylphosphonium Iodide
PubChem CID 2828347
IUPAC Name (N-methylanilino)-triphenylphosphanium;iodide
SMILES CN(C1=CC=CC=C1)[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.[I-]

Specifications

Color White
Quantity 10 g
Formula Weight 495.34
Physical Form Crystalline Powder
Chemical Name or Material (N-Methyl-N-phenylamino)triphenylphosphonium Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.