accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 342573-75-5

IUPAC Name: 1-ethyl-3-methyl-1H-imidazol-3-ium ethyl sulfate
Synonyms: 1-ethyl-3-methylimidazolium ethyl sulfate
Molecular Weight (g/mol): 236.29
Molecular Formula: C8H16N2O4S
InChi Key: VRFOKYHDLYBVAL-UHFFFAOYSA-M
SMILES: CCOS([O-])(=O)=O.CCN1C=C[N+](C)=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)