CAS RN 34286-66-3
IUPAC Name:
2-(2-{[(benzyloxy)carbonyl]amino}propanamido)acetic acid
Synonyms:
(2-{[(benzyloxy)carbonyl]amino}propanamido)acetic acid
Molecular Weight (g/mol):
280.28
Molecular Formula:
C13H16N2O5
InChi Key:
RNBMQRYMCAVZPN-UHFFFAOYNA-N
SMILES:
CC(NC(=O)OCC1=CC=CC=C1)C(=O)NCC(O)=O
1
–
1
of
1
results
1
N-Benzyloxycarbonyl-D-alanylglycine, 95%, Thermo Scientific™
CAS: 34286-66-3 Molecular Formula: C13H16N2O5 Molecular Weight (g/mol): 280.28 MDL Number: MFCD00238424 InChI Key: RNBMQRYMCAVZPN-SECBINFHSA-N Synonym: z-d-ala-gly-oh,r-2-2-benzyloxy carbonyl amino propanamido acetic acid,n-r-2-benzyloxycarbonyl amino-1-oxopropyl glycine,r-2-2-benzyloxy carbonyl amino propanamido aceticacid,2-2r-2-benzyloxy carbonyl amino propanamido acetic acid,2r-2-benzyloxy carbonyl amino propanamido acetic acid,2-2r-2-phenylmethoxycarbonylamino propanoyl amino acetic acid PubChem CID: 7004845 IUPAC Name: 2-[[(2R)-2-(phenylmethoxycarbonylamino)propanoyl]amino]acetic acid SMILES: CC(C(=O)NCC(=O)O)NC(=O)OCC1=CC=CC=C1