Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyloxycarbonyl-D-alanylglycine, 95%, Thermo Scientific™

Catalog No. AAH66802MD
Change view
Click to view available options
Quantity:
250 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66802MD 250 mg
AAH6680206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66802MD Supplier Thermo Scientific Chemicals Supplier No. H66802MD

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34286-66-3
Molecular Formula C13H16N2O5
Molecular Weight (g/mol) 280.28
MDL Number MFCD00238424
InChI Key RNBMQRYMCAVZPN-SECBINFHSA-N
Synonym z-d-ala-gly-oh, r-2-2-benzyloxy carbonyl amino propanamido acetic acid, n-r-2-benzyloxycarbonyl amino-1-oxopropyl glycine, r-2-2-benzyloxy carbonyl amino propanamido aceticacid, 2-2r-2-benzyloxy carbonyl amino propanamido acetic acid, 2r-2-benzyloxy carbonyl amino propanamido acetic acid, 2-2r-2-phenylmethoxycarbonylamino propanoyl amino acetic acid
PubChem CID 7004845
IUPAC Name 2-[[(2R)-2-(phenylmethoxycarbonylamino)propanoyl]amino]acetic acid
SMILES CC(C(=O)NCC(=O)O)NC(=O)OCC1=CC=CC=C1

Specifications

Quantity 250 mg
Formula Weight 280.28
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Benzyloxycarbonyl-D-alanylglycine
storageNote1 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.