CAS RN 342897-54-5
IUPAC Name:
2-methyl-1,3-benzoxazol-4-amine
Synonyms:
2-methyl-1,3-benzoxazol-4-amine
Molecular Weight (g/mol):
148.17
Molecular Formula:
C8H8N2O
InChi Key:
OEULNBJKLTXMSN-UHFFFAOYSA-N
SMILES:
CC1=NC2=C(N)C=CC=C2O1
1
–
1
of
1
results
1
2-Methyl-1,3-benzoxazol-4-amine, 97%, Thermo Scientific™
CAS: 342897-54-5 Molecular Formula: C8H8N2O Molecular Weight (g/mol): 148.165 InChI Key: OEULNBJKLTXMSN-UHFFFAOYSA-N Synonym: 2-methylbenzo d oxazol-4-amine,4-amino-2-methyl-1,3-benzoxazole,methylbenzoxazolamine,2-methyl-4-benzoxazolamine,2-methylbenzoxazol-4-amine,2-methyl-4-aminobenzoxazole,2-methylbenzoxazole-4-amine,4-amino-2-methylbenzoxazole,2-methylbenzoxazole-4-ylamine,4-benzoxazolamine, 2-methyl PubChem CID: 10606897 IUPAC Name: 2-methyl-1,3-benzoxazol-4-amine SMILES: CC1=NC2=C(C=CC=C2O1)N