Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-1,3-benzoxazol-4-amine, 97%, Thermo Scientific™

Catalog No. MAY00305DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAY00305DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAY00305DA Supplier Thermo Scientific Chemicals Supplier No. MAY00305DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 342897-54-5
Molecular Formula C8H8N2O
Molecular Weight (g/mol) 148.165
InChI Key OEULNBJKLTXMSN-UHFFFAOYSA-N
Synonym 2-methylbenzo d oxazol-4-amine, 4-amino-2-methyl-1,3-benzoxazole, methylbenzoxazolamine, 2-methyl-4-benzoxazolamine, 2-methylbenzoxazol-4-amine, 2-methyl-4-aminobenzoxazole, 2-methylbenzoxazole-4-amine, 4-amino-2-methylbenzoxazole, 2-methylbenzoxazole-4-ylamine, 4-benzoxazolamine, 2-methyl
PubChem CID 10606897
IUPAC Name 2-methyl-1,3-benzoxazol-4-amine
SMILES CC1=NC2=C(C=CC=C2O1)N

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 148.16
Chemical Name or Material 2-Methyl-1, 3-benzoxazol-4-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.