accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NBr

CAS RN 3430-16-8

IUPAC Name: 3-bromo-5-methylpyridine
Synonyms: 3-bromo-5-methylpyridine
Molecular Weight (g/mol): 172.03
Molecular Formula: C6H6BrN
InChi Key: ADCLTLQMVAEBLB-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=CN=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)