Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-5-methylpyridine, 97%, Thermo Scientific™

Catalog No. AAH3013703
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3013703 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3013703 Supplier Thermo Scientific Chemicals Supplier No. H3013703

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3430-16-8
Density 1.49
Molecular Formula C6H6BrN
Quantity 1g
MDL Number MFCD01646141
Synonym 5-bromo-3-picoline, 3-bromo-5-picoline, 3-bromo-5-methyl-pyridine, 3-methyl-5-bromopyridine, pyridine, 3-bromo-5-methyl, 5-bromo-3-methylpyridine, 3-brom-5-methylpyridin, zlchem 381, 5-bromo-3-picolin, pubchem4036
InChI Key ADCLTLQMVAEBLB-UHFFFAOYSA-N
SMILES CC1=CC(Br)=CN=C1
IUPAC Name 3-bromo-5-methylpyridine
Molecular Weight (g/mol) 172.03
PubChem CID 817713
Formula Weight 172.03
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-methylpyridine
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 630-960-1
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.