accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3BrNBr

CAS RN 3430-18-0

IUPAC Name: 2,5-dibromo-3-methylpyridine
Synonyms: 2,5-dibromo-3-methylpyridine
Molecular Weight (g/mol): 250.92
Molecular Formula: C6H5Br2N
InChi Key: LIMXEVCFAUTBCK-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=CN=C1Br
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)