Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dibromo-3-methylpyridine 98.0+%, TCI America™
SDP

Catalog No. D34555G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D34555G Supplier TCI America Supplier No. D34555G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 3430-18-0 | C6H5Br2N | 250.92 g/mol

Chemical Identifiers

CAS 3430-18-0
Molecular Formula C6H5Br2N
Molecular Weight (g/mol) 250.92
MDL Number MFCD02093085
InChI Key LIMXEVCFAUTBCK-UHFFFAOYSA-N
Synonym 2,5-dibromo-3-picoline, 2,5-dibromo-3-methyl-pyridine, pyridine, 2,5-dibromo-3-methyl, 2,5-dibromo-3-methylpyridine, 2,5-dibrom-3-methylpyridin, pubchem1138, acmc-209i6e, ksc495g9h, 2,5-dibromo-3-methyl pyridine
PubChem CID 817680
IUPAC Name 2,5-dibromo-3-methylpyridine
SMILES CC1=CC(Br)=CN=C1Br

Specifications

Melting Point 46°C
Color White-Yellow
Boiling Point 114°C
Quantity 5 g
Formula Weight 250.92
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dibromo-3-methylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.