accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNH2

CAS RN 3430-19-1

IUPAC Name: 5-methylpyridin-3-amine
Synonyms: 5-methylpyridin-3-amine
Molecular Weight (g/mol): 108.14
Molecular Formula: C6H8N2
InChi Key: JXUWZXFVCBODAN-UHFFFAOYSA-N
SMILES: CC1=CC(N)=CN=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results