Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-5-methylpyridine 97.0+%, TCI America™
SDP

Catalog No. A23965G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23965G 5 g
A23961G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23965G Supplier TCI America Supplier No. A23965G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3430-19-1
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD04112508
InChI Key JXUWZXFVCBODAN-UHFFFAOYSA-N
Synonym 5-Amino-3-picoline
PubChem CID 2762894
IUPAC Name 5-methylpyridin-3-amine
SMILES CC1=CC(N)=CN=C1

Specifications

Melting Point 65°C
Color White-Yellow
Boiling Point 155°C
Quantity 5 g
UN Number 2811
Formula Weight 108.14
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-5-methylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.