accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH2N

CAS RN 3430-27-1

IUPAC Name: 4-methylpyridin-3-amine
Synonyms: 4-methylpyridin-3-amine
Molecular Weight (g/mol): 108.14
Molecular Formula: C6H8N2
InChi Key: IBKMZYWDWWIWEL-UHFFFAOYSA-N
SMILES: CC1=CC=NC=C1N
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (3)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (4)
  • (2)