Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-4-methylpyridine 98.0+%, TCI America™
SDP

Catalog No. A19575G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19575G 5 g
A195725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A19575G Supplier TCI America Supplier No. A19575G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 3430-27-1 | C6H8N2 | 108.144 g/mol

Chemical Identifiers

CAS 3430-27-1
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00128871
InChI Key IBKMZYWDWWIWEL-UHFFFAOYSA-N
Synonym 3-amino-4-methylpyridine, 3-amino-4-picoline, 3-pyridinamine, 4-methyl, 4-methyl-3-pyridinamine, 4-methyl-pyridin-3-ylamine, unii-9hn55rq07b, 4-methyl-3-pyridylamine, 4-methylpyridine-3-amine, 3-amino-4-methyl pyridine, 3-amino-4-methylpyridine 3-amino-4-picoline
PubChem CID 137935
IUPAC Name 4-methylpyridin-3-amine
SMILES CC1=C(C=NC=C1)N

Specifications

Melting Point 106°C
Boiling Point 254°C
Quantity 5 g
UN Number 2811
Formula Weight 108.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-methylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.