accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3430-40-8

IUPAC Name: 2-(piperidin-1-yl)benzamide
Synonyms: 2-(piperidin-1-yl)benzamide
Molecular Weight (g/mol): 204.27
Molecular Formula: C12H16N2O
InChi Key: VTXYPPVXMJMLCY-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=CC=C1N1CCCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results