accessibility menu, dialog, popup

UserName

Viewing profile as user:
SH

CAS RN 34301-54-7

IUPAC Name: adamantane-1-thiol
Synonyms: adamantane-1-thiol
Molecular Weight (g/mol): 168.3
Molecular Formula: C10H16S
InChi Key: ADJJLNODXLXTIH-UHFFFAOYSA-N
SMILES: SC12CC3CC(CC(C3)C1)C2
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)