accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNH3CCH3NNH3CH2O

CAS RN 34302-69-7

IUPAC Name: bis(2,9-dimethyl-1,10-phenanthroline) hydrate
Synonyms: bis(neocuproine) hydrate
Molecular Weight (g/mol): 434.54
Molecular Formula: C28H26N4O
InChi Key: IEBXFSLFDFHSRD-UHFFFAOYSA-N
SMILES: O.CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1.CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1
Percent Purity 
  • (3)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)