Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neocuproine Hemihydrate 98.0+%, TCI America™
SDP

Catalog No. D07711G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D07711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D07711G Supplier TCI America Supplier No. D07711G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 34302-69-7
Molecular Formula C28H26N4O
Molecular Weight (g/mol) 434.54
MDL Number MFCD00149306,MFCD00004973,MFCD00149306,MFCD23140843
InChI Key IEBXFSLFDFHSRD-UHFFFAOYSA-N
Synonym unii-2sdt9ev86w, 2sdt9ev86w, neocuproine hemihydrate mi, 1,10-phenanthroline, 2,9-dimethyl-, hemihydrate, 1,10-phenanthroline, 2,9-dimethyl-, hydrate 2:1, 2,9-dimethyl-1,10-phenthroline hemihydrate
PubChem CID 67652146
IUPAC Name bis(2,9-dimethyl-1,10-phenanthroline) hydrate
SMILES O.CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1.CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1

Specifications

Melting Point 160°C
Quantity 1 g
Formula Weight 208.26
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Neocuproine Hemihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.